The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178785.3 | NP_848900.2 | 3 - 4 | 744 | 70 |
GenBank mRNA | AK030797.1 | - | 3 - 4 | 747 | 70 |
AK040573.1 | - | 3 - 4 | 732 | 70 | |
AK041479.1 | - | 4 - 5 | 602 | 70 | |
AK088449.1 | - | 3 - 4 | 706 | 70 | |
AK088987.1 | - | 3 - 4 | 722 | 70 | |
AK131128.1 | - | 5 - 6 | 651 | 70 | |
BC132341.1 | - | 5 - 6 | 650 | 70 |