Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
D2hgdh
Entrez Gene ID
98314
Gene Name
D-2-hydroxyglutarate dehydrogenase
Gene Aliases
AA408776, AA408778, AI325464
Chromosome Location
Chr.1: 93824808 - 93852358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK088200.1 - - 377 64
AK088554.1 - - 377 64
AK170226.1 - - 431 64
BC023277.1 - - 398 64
BC117794.2 - 2 - 3 368 64

View Full Product Details