Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gars
Entrez Gene ID
353172
Gene Name
glycyl-tRNA synthetase
Gene Aliases
GENA202, Gena201, Nmf249, Sgrp23
Chromosome Location
Chr.6: 55038001 - 55079504 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180678.3 NP_851009.2 9 - 10 1209 71
GenBank mRNA AK012758.2 - 9 - 10 1204 71
AK146238.1 - 9 - 10 1186 71
AK165857.1 - 9 - 10 1157 71
BC021747.1 - 9 - 10 1190 71

View Full Product Details