Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286040.1 NP_001272969.1 21 - 22 3200 72
NM_001286041.1 NP_001272970.1 20 - 21 3137 72
NM_181413.4 NP_852078.1 21 - 22 3200 72
GenBank mRNA AK080878.1 - 10 - 11 1835 72
AK129094.1 - 20 - 21 3071 72
AK147273.1 - 21 - 22 3135 72
AK160806.1 - 8 - 9 874 72
AK169828.1 - 10 - 11 1146 72
BC010633.1 - 9 - 10 1083 72
BC050847.1 - 21 - 22 3166 72
BC057331.1 - 9 - 10 1344 72

View Full Product Details