Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181413.4 NP_852078.1 23 - 24 3512 55
GenBank mRNA AK080878.1 - 12 - 13 2147 55
AK160806.1 - 10 - 11 1186 55
AK169828.1 - 12 - 13 1458 55
BC010633.1 - 11 - 12 1395 55
BC050847.1 - 23 - 24 3478 55

View Full Product Details