Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181413.4 NP_852078.1 24 - 25 3608 80
GenBank mRNA AK080878.1 - 13 - 14 2244 81
AK169828.1 - 13 - 14 1554 80
BC010633.1 - 12 - 13 1491 80
BC050847.1 - 24 - 25 3574 80

View Full Product Details