Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks1
Entrez Gene ID
224650
Gene Name
ankyrin repeat and SAM domain containing 1
Gene Aliases
Anks1a, mKIAA0229
Chromosome Location
Chr.17: 27909306 - 28062779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286040.1 NP_001272969.1 5 - 6 940 60
NM_001286041.1 NP_001272970.1 4 - 5 877 60
NM_181413.4 NP_852078.1 5 - 6 940 60
GenBank mRNA AK129094.1 - 4 - 5 811 60
AK147273.1 - 5 - 6 875 60
BC050847.1 - 5 - 6 906 60

View Full Product Details