Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rspry1
Entrez Gene ID
67610
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
4930470D19Rik, AI608258
Chromosome Location
Chr.8: 94601941 - 94660277 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026274.4 NP_080550.3 9 - 10 1659 78
GenBank mRNA AK032416.1 - 9 - 10 1622 78
AK076799.1 - 9 - 10 1232 78
AK134544.1 - 6 - 7 526 78
AK141443.1 - 9 - 10 1209 78
AK150941.1 - 8 - 9 1134 78
AK151018.1 - 9 - 10 1629 78
AK152024.1 - 9 - 10 1629 78
AK152435.1 - 8 - 9 1134 78
AK172567.1 - 9 - 10 1625 78
AK173313.1 - 9 - 10 1278 78
BC010833.1 - 9 - 10 1274 78
BC054121.1 - 9 - 10 1645 78

View Full Product Details