Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slamf7
Entrez Gene ID
75345
Gene Name
SLAM family member 7
Gene Aliases
19A, 19A24, 4930560D03Rik, CRACC, CS1
Chromosome Location
Chr.1: 171632403 - 171653037 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144539.5 NP_653122.2 6 - 7 1326 86
GenBank mRNA AB196816.1 - 5 - 6 873 86
AB196817.1 - 5 - 6 873 86
AB196818.1 - 5 - 6 873 86
AB196819.1 - 5 - 6 873 86
AF467909.1 - 5 - 6 873 86
AF467910.1 - 5 - 6 873 86
AK016183.1 - 6 - 7 1125 86
AK030135.1 - 6 - 7 1187 86
AK030148.1 - 6 - 7 1327 86
AK040678.1 - 5 - 6 954 86
AK172665.1 - 5 - 6 920 86

View Full Product Details