Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sowahc
- Entrez Gene ID
-
268301
- Gene Name
- sosondowah ankyrin repeat domain family member C
- Gene Aliases
-
4921515A04Rik, 9430099J10Rik, Ankrd57, C820004L04Rik
- Chromosome Location
-
Chr.10: 59221922 - 59226434 on Build GRCm38
- Gene Symbol
- Sept10
- Entrez Gene ID
-
103080
- Gene Name
- septin 10
- Gene Aliases
-
4921515A04Rik, 9430099J10Rik, AA408298, AI874685
- Chromosome Location
-
Chr.10: 59141627 - 59221847 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK028519.1 |
- |
- |
|
69 |
View Full Product Details