Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ccdc37
- Entrez Gene ID
-
243538
- Gene Name
- coiled-coil domain containing 37
- Gene Aliases
-
AW048498, C030041G11Rik, C230069K22Rik
- Chromosome Location
-
Chr.6: 90403731 - 90430246 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK048790.1 |
- |
1 - 2 |
406 |
106 |
View Full Product Details