The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013813.3 | NP_001013835.2 | - | 2496 | 81 |
NM_173776.3 | NP_776137.2 | 2 - 3 | 400 | 81 | |
GenBank mRNA | AK031222.1 | - | 2 - 3 | 237 | 81 |
AK129448.1 | - | 2 - 3 | 400 | 81 |