The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173778.3 | NP_776139.1 | 3 - 4 | 1040 | 94 |
GenBank mRNA | AK030178.1 | - | 3 - 4 | 1041 | 94 |
BC116842.1 | - | 3 - 4 | 963 | 94 | |
BC116844.1 | - | 3 - 4 | 963 | 94 |