The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173781.4 | NP_776142.1 | 7 - 8 | 643 | 71 |
GenBank mRNA | AB232597.1 | - | 7 - 8 | 560 | 71 |
AK035893.1 | - | 7 - 8 | 602 | 71 | |
AK082732.1 | - | 7 - 8 | 767 | 71 | |
BC059903.1 | - | 3 - 4 | 226 | 71 | |
BC060618.1 | - | 7 - 8 | 643 | 71 |