The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176965.3 | NP_795939.2 | 4 - 5 | 663 | 90 |
GenBank mRNA | AK029790.1 | - | 2 - 3 | 250 | 90 |
BC127154.1 | - | 2 - 3 | 205 | 90 |