Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kirrel
Entrez Gene ID
170643
Gene Name
kin of IRRE like (Drosophila)
Gene Aliases
6720469N11Rik, Kirrel1, Neph1
Chromosome Location
Chr.3: 87078592 - 87174747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170985.1 NP_001164456.1 16 - 16 6242 81
NM_130867.3 NP_570937.2 16 - 16 6239 81
GenBank mRNA AK032898.1 - 1 - 1 2821 81
AK034569.1 - 1 - 1 1451 81
BC062651.1 - 1 - 1 1284 81

View Full Product Details