The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177304.3 | NP_796278.1 | 3 - 4 | 582 | 72 |
GenBank mRNA | AK038460.1 | - | 3 - 4 | 554 | 72 |
AK046797.1 | - | 3 - 4 | 583 | 72 | |
AK046881.1 | - | 3 - 4 | 587 | 72 | |
AK079352.1 | - | 3 - 4 | 564 | 72 | |
AK079642.1 | - | 3 - 4 | 582 | 72 | |
BC096376.1 | - | 3 - 4 | 553 | 72 |