The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177307.3 | NP_796281.1 | 4 - 5 | 546 | 104 |
GenBank mRNA | AK028950.1 | - | 4 - 5 | 547 | 104 |
AK037192.1 | - | 4 - 5 | 547 | 104 | |
BC145755.1 | - | 4 - 5 | 543 | 104 |