Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cndp1
Entrez Gene ID
338403
Gene Name
carnosine dipeptidase 1 (metallopeptidase M20 family)
Gene Aliases
AI746433, Cn1
Chromosome Location
Chr.18: 84610384 - 84650113 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177450.4 NP_803233.2 1 - 2 195 64
GenBank mRNA AK085308.1 - 1 - 2 185 64
AK165500.1 - 1 - 2 46 64
AK165706.1 - 1 - 2 147 64
BC043305.1 - 1 - 2 124 64
BY006586.1 - 1 - 2 64

View Full Product Details