The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177688.4 | NP_808356.1 | 1 - 1 | 1417 | 95 |
GenBank mRNA | AK053755.1 | - | 1 - 1 | 1344 | 95 |