The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177693.3 | NP_808361.1 | 1 - 2 | 55 | 58 |
GenBank mRNA | AK029354.1 | - | 1 - 2 | 56 | 58 |
AK164958.1 | - | 1 - 2 | 54 | 58 | |
BC092386.1 | - | 1 - 2 | 68 | 58 | |
BC119253.1 | - | 1 - 2 | 36 | 58 | |
BC119255.1 | - | 1 - 2 | 36 | 58 |