The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027626.1 | NP_081902.1 | 6 - 7 | 959 | 63 |
NM_030263.5 | NP_084539.2 | 11 - 12 | 1301 | 63 | |
NM_177698.4 | NP_808366.2 | 14 - 15 | 3065 | 63 | |
GenBank mRNA | AB220685.1 | - | 14 - 15 | 3065 | 63 |
AK031477.1 | - | 6 - 7 | 902 | 63 | |
AK147606.1 | - | 6 - 7 | 959 | 63 | |
AK161498.1 | - | 11 - 12 | 1287 | 63 | |
BC042208.1 | - | 2 - 3 | 288 | 63 | |
BC052365.1 | - | 11 - 12 | 1266 | 63 | |
BC131913.1 | - | 6 - 7 | 871 | 63 | |
BC145641.1 | - | 6 - 7 | 871 | 63 |