Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eme1
Entrez Gene ID
268465
Gene Name
essential meiotic endonuclease 1 homolog 1 (S. pombe)
Gene Aliases
6820428D13
Chromosome Location
Chr.11: 94644998 - 94653785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177752.4 NP_808420.1 4 - 5 1078 60
GenBank mRNA AK078516.1 - 4 - 5 1079 60
BC089459.1 - 4 - 5 1053 60

View Full Product Details