The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177765.3 | NP_808433.2 | 14 - 15 | 2516 | 71 |
GenBank mRNA | AK029565.1 | - | 14 - 15 | 2515 | 71 |
AM690757.1 | - | 13 - 14 | 2217 | 71 |