Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc16a
Entrez Gene ID
68732
Gene Name
leucine rich repeat containing 16A
Gene Aliases
1110037D04Rik, AI425970, CARMIL, D130057M20, Lrrc16
Chromosome Location
Chr.13: 24012481 - 24280790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026825.3 NP_081101.3 3 - 4 377 93
GenBank mRNA AK051570.1 - 3 - 4 377 93
AY437876.1 - 3 - 4 382 93
BC150819.1 - 3 - 4 303 93

View Full Product Details