The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177900.4 | NP_808568.1 | 4 - 5 | 841 | 64 |
GenBank mRNA | AB107882.1 | - | 4 - 5 | 841 | 64 |
AK034300.1 | - | 4 - 5 | 850 | 64 | |
AY269789.1 | - | 4 - 5 | 913 | 64 | |
BC125339.1 | - | 4 - 5 | 828 | 64 | |
BC125341.1 | - | 4 - 5 | 828 | 64 |