The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177909.5 | NP_808577.3 | 14 - 15 | 1777 | 69 |
GenBank mRNA | AK037058.1 | - | 14 - 15 | 1761 | 69 |
AK156494.1 | - | 14 - 15 | 1738 | 69 | |
AK172687.1 | - | 14 - 15 | 1749 | 69 | |
BC099942.1 | - | 14 - 15 | 1751 | 69 | |
BC153825.1 | - | 14 - 15 | 1755 | 69 |