Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pramel6
Entrez Gene ID
347711
Gene Name
preferentially expressed antigen in melanoma like 6
Gene Aliases
C80453, C80515
Chromosome Location
Chr.2: 87506565 - 87510868 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178249.2 NP_839980.1 1 - 2 284 82
GenBank mRNA AF490342.1 - 1 - 2 284 82
AK139830.1 - 2 - 3 518 82

View Full Product Details