Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pkd2
Entrez Gene ID
18764
Gene Name
polycystic kidney disease 2
Gene Aliases
C030034P18Rik, PC2, TRPP2
Chromosome Location
Chr.5: 104457224 - 104505819 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF014010.1 - 12 - 13 2434 78
AK128961.1 - 13 - 14 2305 78

View Full Product Details