Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyb5b
Entrez Gene ID
66427
Gene Name
cytochrome b5 type B
Gene Aliases
1810044O22Rik, AU015618, Cyb5m
Chromosome Location
Chr.8: 107150661 - 107187470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025558.5 NP_079834.2 1 - 2 213 75
GenBank mRNA AK003333.2 - 1 - 2 214 75
AK003815.1 - 1 - 2 203 75
AK007780.1 - - 233 75
AK019263.1 - 1 - 2 213 75
AK052810.1 - 1 - 2 214 75
AK088970.1 - 1 - 2 215 75
AK165543.1 - 1 - 2 217 75
AK166544.1 - 1 - 2 191 75
AK168422.1 - 1 - 2 203 75
BC046479.1 - 1 - 2 176 75
BC046810.1 - 1 - 2 179 75
BC048147.1 - 1 - 2 203 75
BC054749.1 - 1 - 2 234 75
BC058812.1 - 1 - 2 176 75
BC062980.1 - 1 - 2 225 75

View Full Product Details