The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172476.4 | NP_766064.2 | 14 - 15 | 2047 | 60 |
GenBank mRNA | AK083189.1 | - | 14 - 15 | 2050 | 60 |
AK132820.1 | - | 14 - 15 | 2079 | 60 | |
AY236499.1 | - | 14 - 15 | 2108 | 60 | |
AY263161.1 | - | 14 - 15 | 2076 | 60 | |
BC105966.1 | - | 8 - 9 | 1061 | 60 |