Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt15
Entrez Gene ID
319508
Gene Name
synaptotagmin XV
Gene Aliases
CHR10SYT, E230025K04Rik, sytXV
Chromosome Location
Chr.14: 34220046 - 34230421 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176931.2 NP_795905.2 3 - 4 477 85
NM_181529.4 NP_852682.1 3 - 4 477 85
GenBank mRNA AB109019.1 - 2 - 3 349 85
AB109020.1 - 2 - 3 349 85
AK042641.1 - 3 - 4 478 85
AK054180.1 - 3 - 4 477 85
BB639494.1 - 3 - 4 478 85
BC139313.1 - 3 - 4 388 85
BC139314.1 - 3 - 4 389 85

View Full Product Details