The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029385.2 | NP_083661.2 | 2 - 3 | 435 | 74 |
GenBank mRNA | AK009737.1 | - | 2 - 3 | 436 | 74 |
AK012920.1 | - | 1 - 2 | 488 | 74 | |
BC064048.1 | - | 2 - 3 | 422 | 74 |