The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028232.2 | NP_082508.1 | 5 - 6 | 659 | 127 |
GenBank mRNA | AB193067.1 | - | 4 - 5 | 567 | 127 |
AB193068.1 | - | 4 - 5 | 567 | 127 | |
AK014357.1 | - | 5 - 6 | 653 | 127 | |
AK154188.1 | - | 5 - 6 | 678 | 127 | |
BC089014.1 | - | 4 - 5 | 506 | 127 |