Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r80
Entrez Gene ID
171238
Gene Name
vomeronasal 1 receptor 80
Gene Aliases
V1rg3
Chromosome Location
Chr.7: 12192965 - 12193891 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134204.2 NP_598965.2 1 - 1 852 128

View Full Product Details