Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcbld2
Entrez Gene ID
73379
Gene Name
discoidin, CUB and LCCL domain containing 2
Gene Aliases
1700055P21Rik, AW146002, CLCP1, Esdn
Chromosome Location
Chr.16: 58408426 - 58469745 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028523.3 NP_082799.2 9 - 10 1422 114
GenBank mRNA AF387548.1 - 9 - 10 1514 114
AK051889.1 - 7 - 8 700 114
BC066097.1 - 9 - 10 1422 114

View Full Product Details