The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146934.2 | NP_667145.1 | 3 - 3 | 690 | 102 |
GenBank mRNA | AF102520.1 | - | 1 - 1 | 244 | 102 |
AF102526.1 | - | 1 - 1 | 244 | 102 | |
AF102531.1 | - | - | 244 | 102 | |
BC108947.1 | - | 1 - 1 | 455 | 102 |