The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177150.2 | NP_796124.1 | 10 - 11 | 1164 | 89 |
GenBank mRNA | AK089172.1 | - | 10 - 11 | 1213 | 89 |
AK090340.1 | - | 10 - 11 | 1165 | 89 | |
AK143613.1 | - | 10 - 11 | 1127 | 89 | |
AK164039.1 | - | 6 - 7 | 765 | 89 | |
AK167376.1 | - | 10 - 11 | 1165 | 89 | |
BC121824.1 | - | 10 - 11 | 1165 | 89 |