The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112739.2 | NP_001106210.1 | 1 - 2 | 1785 | 128 |
NM_008421.3 | NP_032447.1 | 1 - 2 | 1785 | 128 | |
GenBank mRNA | AK147476.1 | - | 1 - 2 | 1758 | 128 |
AK147682.1 | - | 1 - 2 | 1758 | 128 | |
AK162964.1 | - | 1 - 2 | 442 | 128 | |
BC125470.1 | - | 1 - 2 | 622 | 128 | |
BC132439.1 | - | 1 - 2 | 622 | 128 | |
Y07521.1 | - | 1 - 2 | 711 | 128 |