The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256005.1 | NP_001242934.1 | 6 - 7 | 1024 | 144 |
NM_008620.3 | NP_032646.2 | 6 - 7 | 961 | 144 | |
GenBank mRNA | AK220567.1 | - | 6 - 7 | 942 | 144 |
BC141364.1 | - | 6 - 7 | 912 | 144 | |
BC147434.1 | - | 6 - 7 | 912 | 144 | |
EF494423.1 | - | 6 - 7 | 1024 | 144 | |
EF494424.1 | - | 6 - 7 | 995 | 144 |