The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008648.1 | NP_032674.1 | 5 - 6 | 575 | 160 |
GenBank mRNA | BC012227.1 | - | 5 - 6 | 562 | 160 |
M16358.1 | - | 5 - 6 | 575 | 160 |