The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010643.1 | NP_034773.1 | - | 646 | 161 |
GenBank mRNA | AB039277.1 | - | - | 646 | 161 |
BC146015.1 | - | 4 - 5 | 673 | 161 | |
BC146019.1 | - | 4 - 5 | 673 | 161 |