The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290759.1 | NP_001277688.1 | 21 - 22 | 3162 | 82 |
NM_001290760.1 | NP_001277689.1 | 21 - 22 | 3165 | 82 | |
NM_029745.2 | NP_084021.2 | 20 - 21 | 3114 | 82 | |
GenBank mRNA | AK031992.1 | - | 6 - 7 | 584 | 82 |
AK035457.1 | - | 20 - 21 | 3079 | 82 | |
AK088860.1 | - | 15 - 16 | 2064 | 82 | |
AK147964.1 | - | 20 - 21 | 3079 | 82 | |
AK172999.1 | - | 22 - 23 | 3102 | 82 | |
AK184661.1 | - | 4 - 5 | 255 | 82 | |
AK194602.1 | - | 4 - 5 | 256 | 82 | |
AK207865.1 | - | 2 - 3 | 147 | 82 | |
AK207874.1 | - | 3 - 4 | 204 | 82 | |
AK210062.1 | - | 2 - 3 | 83 | ||
AK210947.1 | - | 2 - 3 | 147 | 82 | |
AK212277.1 | - | 4 - 5 | 255 | 82 | |
AK219161.1 | - | 2 - 3 | 147 | 82 | |
BC048085.1 | - | 11 - 12 | 1428 | 82 | |
BC058596.1 | - | 8 - 9 | 699 | 82 | |
BC062928.1 | - | 20 - 21 | 3101 | 82 | |
BC065080.1 | - | 20 - 21 | 3093 | 82 | |
BC065130.1 | - | 20 - 21 | 3057 | 82 |