The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011702.2 | NP_035832.1 | 5 - 6 | 634 | 82 |
GenBank mRNA | AK018599.1 | - | 5 - 6 | 635 | 82 |
AK043544.1 | - | 4 - 5 | 2355 | 82 | |
BC089511.1 | - | 5 - 6 | 540 | 82 |