Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc16a
Entrez Gene ID
68732
Gene Name
leucine rich repeat containing 16A
Gene Aliases
1110037D04Rik, AI425970, CARMIL, D130057M20, Lrrc16
Chromosome Location
Chr.13: 24012481 - 24280790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026825.3 NP_081101.3 25 - 26 2268 89
GenBank mRNA AK043591.1 - 10 - 11 848 89
AK049059.1 - 7 - 8 644 89
AY437876.1 - 25 - 26 2273 89
BC012229.1 - 4 - 5 297 89
BC150819.1 - 24 - 25 2182 89

View Full Product Details