The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026879.2 | NP_081155.1 | 4 - 5 | 617 | 129 |
GenBank mRNA | AK004506.1 | - | 1 - 2 | 90 | 129 |
AK084111.1 | - | 4 - 5 | 619 | 129 | |
BC042626.1 | - | 3 - 4 | 373 | 129 | |
BC055809.1 | - | 4 - 5 | 646 | 129 |