The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081309.1 | NP_001074778.1 | 4 - 5 | 1890 | 97 |
GenBank mRNA | AK042953.1 | - | 4 - 5 | 1878 | 97 |
AK050333.1 | - | 4 - 5 | 1860 | 97 | |
AK054559.1 | - | 4 - 5 | 1907 | 97 | |
AK087938.1 | - | 4 - 5 | 1881 | 97 | |
AK169508.1 | - | 3 - 4 | 2677 | 97 | |
BC017537.1 | - | 3 - 4 | 1492 | 97 | |
BC024083.1 | - | 1 - 2 | 364 | 97 | |
BC157947.1 | - | 4 - 5 | 1715 | 97 | |
BC158047.1 | - | 4 - 5 | 1715 | 97 |