Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd55
Entrez Gene ID
77318
Gene Name
ankyrin repeat domain 55
Gene Aliases
C030011J08Rik
Chromosome Location
Chr.13: 112288451 - 112384002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168404.1 NP_001161876.1 2 - 3 171 65
NM_001168405.1 NP_001161877.1 1 - 2 528 65
NM_029898.3 NP_084174.2 1 - 2 528 65
GenBank mRNA AK039553.1 - 1 - 2 529 65
AK045480.1 - 1 - 2 162 65
AK081197.1 - 2 - 3 172 65
AK138273.1 - 1 - 2 183 65
BC152317.1 - 1 - 2 155 65
BC152536.1 - 1 - 2 155 65

View Full Product Details