Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd55
Entrez Gene ID
77318
Gene Name
ankyrin repeat domain 55
Gene Aliases
C030011J08Rik
Chromosome Location
Chr.13: 112288451 - 112384002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168403.1 NP_001161875.1 10 - 11 2212 110
NM_029898.3 NP_084174.2 9 - 10 2106 110
GenBank mRNA AK021066.1 - 1 - 2 257 110
AK045480.1 - 9 - 10 1740 110
AK140417.1 - 10 - 11 2076 110
BC152317.1 - 9 - 10 1733 110
BC152536.1 - 9 - 10 1733 110

View Full Product Details