Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Dgkz
- Entrez Gene ID
-
104418
- Gene Name
- diacylglycerol kinase zeta
- Gene Aliases
-
E130307B02Rik, ENSMUSG00000075035, F730209L11Rik, mDGK[z]
- Chromosome Location
-
Chr.2: 91932822 - 91976443 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK088841.1 |
- |
|
|
|
Chr. - - [-] on Build |
61 |
View Full Product Details